siRNA Details
Gene Symbol: Fbxl7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108545.1 | NP_001102015.1 | 4 | 1667 | ||
XM_006232087.2 | XP_006232149.1 | 3 | 3690 | |||
XM_008760775.1 | XP_008758997.1 | 4 | 1459 | |||
XM_008760776.1 | XP_008758998.1 | 5 | 1363 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
