siRNA Details
Gene Symbol: Ect2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108547.2 | NP_001102017.2 | 20 | 2149 | ||
GenBank mRNA | BC168962.1 | - | 3 | 123 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
