siRNA Details
Gene Symbol: Ect2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108547.2 | NP_001102017.2 | 7 | 750 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001177626, NM_001177625, NM_007900, BC023881, BC025565, and others.
Gene Ontology Categories:
