siRNA Details
Gene Symbol: Bbs7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012180.1 | NP_001012180.1 | 16, 17 | 1883 | ||
GenBank mRNA | BC081968.1 | - | 16, 17 | 1883 | 7125501 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_027810, AF521645, AK132661, BC031505.
Gene Ontology Categories:
