siRNA Details
Gene Symbol: Mfsd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047910.1 | NP_001041375.1 | 6 | 688 | ||
GenBank mRNA | AY325200.1 | - | 6 | 688 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mfsd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047910.1 | NP_001041375.1 | 6 | 688 | ||
GenBank mRNA | AY325200.1 | - | 6 | 688 |