siRNA Details
Gene Symbol: RGD1359508 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009707.1 | NP_001009707.1 | 6 | 464 | ||
XM_006232306.2 | XP_006232368.1 | 6 | 740 | |||
XM_006232307.2 | XP_006232369.1 | 5 | 435 | |||
XM_006232308.2 | XP_006232370.1 | 6 | 526 | |||
GenBank mRNA | BC088236.1 | - | 6 | 464 | 7325191 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
