siRNA Details
Gene Symbol: Postn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108550.1 | NP_001102020.1 | 7 | 857 | ||
XM_006232335.2 | XP_006232397.1 | 7 | 884 | |||
XM_006232336.2 | XP_006232398.1 | 7 | 883 | |||
XM_006232337.2 | XP_006232399.1 | 7 | 882 | |||
GenBank mRNA | KM117173.1 | - | 7 | 839 |
