siRNA Details
Gene Symbol: Postn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108550.1 | NP_001102020.1 | 17, 18 | 2121 | ||
XM_006232335.2 | XP_006232397.1 | 17, 18 | 2148 |

More Information
Other Species Targeted

Gene Ontology Categories:
