siRNA Details
Gene Symbol: RGD1562629 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008761027.1 | XP_008759249.1 | 15, 16 | 2301 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_030595, Y18276.
Gene Ontology Categories:
