siRNA Details
Gene Symbol: Mfsd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191847.1 | NP_001178776.1 | 8, 9 | 780 | ||
XM_008761056.1 | XP_008759278.1 | 8, 9 | 782 | |||
GenBank mRNA | FQ211604.1 | - | 797 | |||
FQ217493.1 | - | 765 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
