siRNA Details
Gene Symbol: Zbbx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029922.1 | NP_001025093.1 | 6 | 564 | ||
XM_006232495.2 | XP_006232557.1 | 6 | 480 | |||
XM_006232496.2 | XP_006232558.1 | 6 | 479 | |||
XM_008761103.1 | XP_008759325.1 | 8 | 927 | |||
XM_008761104.1 | XP_008759326.1 | 8 | 926 | |||
XM_008761105.1 | XP_008759327.1 | 8 | 930 | |||
GenBank mRNA | BC097393.1 | - | 6 | 564 | 7453249 |
