siRNA Details
Gene Symbol: Zbbx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029922.1 | NP_001025093.1 | 9, 10 | 1031 | ||
XM_006232495.2 | XP_006232557.1 | 9, 10 | 962 | |||
XM_006232496.2 | XP_006232558.1 | 9, 10 | 961 | |||
XM_008761103.1 | XP_008759325.1 | 11, 12 | 1409 | |||
XM_008761104.1 | XP_008759326.1 | 11, 12 | 1393 | |||
XM_008761105.1 | XP_008759327.1 | 11, 12 | 1412 | |||
GenBank mRNA | BC097393.1 | - | 9, 10 | 1031 | 7453249 |
