siRNA Details
Gene Symbol: Tmem144 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108551.1 | NP_001102021.1 | 6 | 788 | ||
XM_006232517.2 | XP_006232579.1 | 6 | 789 | |||
GenBank mRNA | FQ211169.1 | - | 4 | 342 | ||
FQ221228.1 | - | 4 | 342 | |||
FQ223804.1 | - | 4 | 342 |
