siRNA Details
Gene Symbol: Fga | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008724.1 | NP_001008724.1 | 6 | 1249 | ||
NM_052797.1 | NP_434684.1 | 5 | 1249 | |||
XM_006232532.2 | XP_006232594.1 | 5 | 1293 | |||
XM_006232533.2 | XP_006232595.1 | 5 | 1285 | |||
GenBank mRNA | AY310161.1 | - | 6 | 1249 | ||
BC098766.1 | - | 1 | 434 | |||
BC127465.1 | - | 5 | 1249 | 8375711 | ||
FQ209852.1 | - | 5 | 1267 |
