siRNA Details
Gene Symbol: Rrnad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014173.1 | NP_001014195.1 | 7, 8 | 1543 | ||
XM_006232686.2 | XP_006232748.1 | 6, 7 | 1375 | |||
XM_006232688.2 | XP_006232750.1 | 6, 7 | 1100 | |||
XM_008761179.1 | XP_008759401.1 | 6, 7 | 1335 | |||
GenBank mRNA | BC079059.1 | - | 7, 8 | 1543 | 7115706 |

More Information
Other Species Targeted

