siRNA Details
Gene Symbol: RGD1565775 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_342277.6 | XP_342278.5 | 12 | 1498 | ||
GenBank mRNA | BC092205.1 | - | 12 | 1451 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_028814, AK012969, AK218535, BC006621, AK170490.
Gene Ontology Categories:
