siRNA Details
Gene Symbol: LOC361985 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014174.1 | NP_001014196.1 | 2, 3 | 352 | ||
GenBank mRNA | BC083693.1 | - | 2, 3 | 352 | 7189519 | |
BC098673.1 | - | 2, 3 | 334 | 7130789 | ||
BC105817.1 | - | 2, 3 | 336 | 7368048 | ||
FQ214790.1 | - | 2, 3 | 369 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
