siRNA Details
Gene Symbol: Chtop | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014175.1 | NP_001014197.1 | 5 | 812 | ||
XM_006232710.2 | XP_006232772.1 | 6 | 1780 | |||
XM_006232711.2 | XP_006232773.1 | 6 | 1696 | |||
XM_006232713.2 | XP_006232775.1 | 6 | 1692 | |||
XM_006232715.2 | XP_006232777.1 | 5 | 1558 | |||
GenBank mRNA | BC081985.1 | - | 5 | 812 | 7128951 | |
BC100084.1 | - | 6 | 728 | 7385093 | ||
FQ231784.1 | - | 5 | 630 |
Gene Symbol: Chtop | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749811.3 | XP_003749859.2 | 1 | 925 | ||
XM_006237107.2 | XP_006237169.1 | 2 | 787 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
