siRNA Details
Gene Symbol: Chtop | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014175.1 | NP_001014197.1 | 3 | 482 | ||
XM_006232710.2 | XP_006232772.1 | 3 | 1309 | |||
XM_006232711.2 | XP_006232773.1 | 3 | 1225 | |||
XM_006232713.2 | XP_006232775.1 | 3 | 1224 | |||
XM_006232715.2 | XP_006232777.1 | 3 | 1225 | |||
GenBank mRNA | BC081985.1 | - | 3 | 482 | 7128951 | |
BC100084.1 | - | 3 | 260 | 7385093 | ||
FQ231784.1 | - | 2 | 162 |
Gene Symbol: Chtop | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749811.3 | XP_003749859.2 | 1 | 454 | ||
XM_006237107.2 | XP_006237169.1 | 1 | 454 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_023215, BC144775, BC120651, AK135914, AK047914, and others.
Gene Ontology Categories:
