siRNA Details
Gene Symbol: S100a16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108557.2 | NP_001102027.1 | 2 | 151 | ||
XM_006232717.2 | XP_006232779.1 | 2 | 225 | |||
XM_006232718.1 | XP_006232780.1 | 2 | 142 | |||
GenBank mRNA | BC158598.1 | - | 2 | 151 | 7127572 | |
FQ209774.1 | - | 2 | 153 | |||
FQ214291.1 | - | 2 | 134 | |||
FQ214430.1 | - | 2 | 147 | |||
FQ229313.1 | - | 2 | 159 |
