siRNA Details
Gene Symbol: S100a16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108557.2 | NP_001102027.1 | 3 | 306 | ||
XM_006232717.2 | XP_006232779.1 | 3 | 380 | |||
XM_006232718.1 | XP_006232780.1 | 3 | 297 | |||
GenBank mRNA | BC158598.1 | - | 3 | 306 | 7127572 | |
FQ209774.1 | - | 3 | 308 | |||
FQ214291.1 | - | 3 | 289 | |||
FQ214430.1 | - | 3 | 302 | |||
FQ229313.1 | - | 3 | 314 |
