siRNA Details
Gene Symbol: Prpf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108559.1 | NP_001102029.1 | 7 | 1174 | ||
XM_006232968.2 | XP_006233030.1 | 7 | 1182 | |||
XM_006232969.2 | XP_006233031.1 | 7 | 2856 | |||
XM_006232970.1 | XP_006233032.1 | 7 | 1148 | |||
XM_008761313.1 | XP_008759535.1 | 7 | 1183 | |||
XM_008761314.1 | XP_008759536.1 | 7 | 1184 |
