siRNA Details
Gene Symbol: Anp32e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013200.1 | NP_001013218.1 | 2, 3 | 472 | ||
GenBank mRNA | BC083744.1 | - | 2, 3 | 472 | 7192147 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
