siRNA Details
Gene Symbol: Ampd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101681.2 | NP_001095151.1 | 11 | 1336 | ||
XM_006233168.1 | XP_006233230.1 | 12 | 1516 | |||
XM_006233169.2 | XP_006233231.1 | 12 | 1486 | |||
XM_006233170.2 | XP_006233232.1 | 11 | 1436 | |||
XM_008761407.1 | XP_008759629.1 | 10 | 1304 | |||
GenBank mRNA | BC166402.1 | - | 11 | 1336 | 9028857 |

More Information
Other Species Targeted

Gene Ontology Categories:
