siRNA Details
Gene Symbol: Alg14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014176.2 | NP_001014198.1 | 2 | 318 | ||
XM_006233243.2 | XP_006233305.1 | 2 | 349 | |||
XM_008761465.1 | XP_008759687.1 | 2 | 349 | |||
XM_008761466.1 | XP_008759688.1 | 2 | 348 | |||
GenBank mRNA | BC079151.1 | - | 2 | 264 | 7128610 | |
CO400199.1 | - | 2 | 318 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
