siRNA Details
Gene Symbol: Ank2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008761478.1 | XP_008759700.1 | 20, 21 | 6679 | ||
GenBank mRNA | U65916.1 | - | 21, 22 | 2374 |

More Information
Other Species Targeted

Gene Ontology Categories:
