siRNA Details
Gene Symbol: Ap1ar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191850.1 | NP_001178779.1 | 9 | 565 | ||
XM_006233291.2 | XP_006233353.2 | 9 | 898 | |||
XM_006233292.2 | XP_006233354.2 | 8 | 799 | |||
GenBank mRNA | BC091336.1 | - | 2 | 113 |
