siRNA Details
Gene Symbol: Lamtor3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008375.1 | NP_001008376.1 | 6, 7 | 426 | ||
XM_006233383.2 | XP_006233445.1 | 6, 7 | 551 | |||
XM_006233385.2 | XP_006233447.1 | 6, 7 | 496 | |||
GenBank mRNA | BC086353.1 | - | 6, 7 | 426 | 7304027 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_019920, BC016417, AK151432, AK162102, BC083155, and others.
Gene Ontology Categories:
