siRNA Details
Gene Symbol: Gbp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108569.3 | NP_001102039.2 | 5, 6 | 866 | ||
GenBank mRNA | BC168951.1 | - | 1, 2 | 78 | ||
CO559037.1 | - | 3, 4 | 532 | |||
CO559828.1 | - | 1, 2 | 77 |
