siRNA Details
Gene Symbol: Gbp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108569.3 | NP_001102039.2 | 7, 8 | 1389 | ||
GenBank mRNA | BC168951.1 | - | 3, 4 | 601 | ||
CO559828.1 | - | 3, 4 | 600 |

More Information
Other Species Targeted

Gene Ontology Categories:
