siRNA Details
Gene Symbol: Gbp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108569.3 | NP_001102039.2 | 7 | 1370 | ||
GenBank mRNA | BC168951.1 | - | 3 | 582 | ||
CO559828.1 | - | 3 | 581 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gbp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108569.3 | NP_001102039.2 | 7 | 1370 | ||
GenBank mRNA | BC168951.1 | - | 3 | 582 | ||
CO559828.1 | - | 3 | 581 |