siRNA Details
Gene Symbol: Il36b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006233614.1 | XP_006233676.1 | 1 | 72 | ||
XM_006233615.1 | XP_006233677.1 | 2 | 1392 | |||
XM_006233616.2 | XP_006233678.1 | 2 | 275 | |||
XM_006233617.2 | XP_006233679.1 | 2 | 488 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
