siRNA Details
Gene Symbol: Snapc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749426.3 | XP_003749474.1 | 7, 8 | 2089 | ||
XM_006233716.2 | XP_006233778.1 | 7, 8 | 2085 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Snapc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749426.3 | XP_003749474.1 | 7, 8 | 2089 | ||
XM_006233716.2 | XP_006233778.1 | 7, 8 | 2085 |