siRNA Details
Gene Symbol: LOC102554283 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006233729.2 | XP_006233791.1 | 2 | 834 |
Gene Symbol: LOC102554393 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006233879.2 | XP_006233941.2 | 17 | 2066 |

More Information
Other Species Targeted

Gene Ontology Categories:
