siRNA Details
Gene Symbol: Endog | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034938.1 | NP_001030110.1 | 2 | 570 | ||
GenBank mRNA | AB221075.1 | - | 2 | 570 | ||
FQ232745.1 | - | 2 | 620 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
