siRNA Details
Gene Symbol: Fubp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039337.1 | NP_001034426.1 | 15 | 1393 | ||
XM_006233921.2 | XP_006233983.1 | 15 | 1538 | |||
XM_006233922.2 | XP_006233984.1 | 15 | 1538 | |||
XM_006233924.2 | XP_006233986.1 | 15 | 1539 | |||
GenBank mRNA | DQ144645.1 | - | 15 | 1393 | ||
FQ212354.1 | - | 15 | 1471 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
