siRNA Details
Gene Symbol: Fubp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039337.1 | NP_001034426.1 | 8 | 646 | ||
XM_006233921.2 | XP_006233983.1 | 8 | 791 | |||
XM_006233922.2 | XP_006233984.1 | 8 | 791 | |||
XM_006233924.2 | XP_006233986.1 | 8 | 792 | |||
GenBank mRNA | DQ144645.1 | - | 8 | 646 | ||
FQ212354.1 | - | 8 | 724 |
