siRNA Details
Gene Symbol: Ptrh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749449.3 | XP_003749497.2 | 4 | 552 | ||
GenBank mRNA | FQ220792.1 | - | 4 | 437 | ||
FQ229591.1 | - | 2 | 60 |

More Information
Other Species Targeted

Gene Ontology Categories:
