siRNA Details
Gene Symbol: Fam129b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109885.1 | NP_001103355.1 | 3 | 414 | ||
GenBank mRNA | BC168248.1 | - | 3 | 336 | 5598117 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fam129b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109885.1 | NP_001103355.1 | 3 | 414 | ||
GenBank mRNA | BC168248.1 | - | 3 | 336 | 5598117 |