siRNA Details
Gene Symbol: RGD1308742 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006234051.2 | XP_006234113.2 | 8 | 854 | ||
XM_008761799.1 | XP_008760021.1 | 6 | 620 | |||
XM_008761800.1 | XP_008760022.1 | 6 | 617 | |||
XM_342422.5 | XP_342423.5 | 8 | 851 |
