siRNA Details
Gene Symbol: Nr6a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008761807.1 | XP_008760029.1 | 10 | 1647 | ||
GenBank mRNA | AJ783965.1 | - | 1 | 51 |

More Information
Other Species Targeted

Gene Ontology Categories:
