siRNA Details
Gene Symbol: Nr6a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008761807.1 | XP_008760029.1 | 8, 9 | 1476 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Nr6a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008761807.1 | XP_008760029.1 | 8, 9 | 1476 |