siRNA Details
Gene Symbol: Lypd6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134580.1 | NP_001128052.1 | 9 | 787 | ||
XM_008761762.1 | XP_008759984.1 | 6 | 2633 | |||
XM_008761763.1 | XP_008759985.1 | 7 | 3283 | |||
XM_008761764.1 | XP_008759986.1 | 6 | 3099 | |||
XM_008761765.1 | XP_008759987.1 | 8 | 3403 | |||
XM_008761766.1 | XP_008759988.1 | 5 | 2573 | |||
XM_008761767.1 | XP_008759989.1 | 5 | 3039 | |||
XM_008761768.1 | XP_008759990.1 | 5 | 2083 | |||
XM_008761769.1 | XP_008759991.1 | 6 | 2539 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
