siRNA Details
Gene Symbol: Hnrnpa3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111294.1 | NP_001104764.1 | 4, 5 | 633 | ||
NM_001111295.1 | NP_001104765.1 | 4, 5 | 567 | |||
NM_198132.3 | NP_937765.1 | 4, 5 | 633 | |||
XM_008761911.1 | XP_008760133.1 | 4, 5 | 582 | |||
XM_008761912.1 | XP_008760134.1 | 4, 5 | 582 | |||
XM_008761913.1 | XP_008760135.1 | 4, 5 | 628 | |||
GenBank mRNA | AY363226.1 | - | 4, 5 | 579 | ||
AY363227.1 | - | 4, 5 | 513 | |||
BC081878.1 | - | 4, 5 | 573 | 7108128 | ||
CB725005.1 | - | 4, 5 | 505 | |||
FQ214617.1 | - | 4, 5 | 529 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_146130, NM_198090, NM_053263, BC023908, BC157904, and others.
Gene Ontology Categories:
