siRNA Details
Gene Symbol: Zdhhc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039338.1 | NP_001034427.1 | 10 | 1358 | ||
GenBank mRNA | AY871203.1 | - | 10 | 1358 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zdhhc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039338.1 | NP_001034427.1 | 10 | 1358 | ||
GenBank mRNA | AY871203.1 | - | 10 | 1358 |