siRNA Details
Gene Symbol: Celf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025421.1 | NP_001020592.1 | 10 | 1155 | ||
XM_006234513.2 | XP_006234575.1 | 12 | 2086 | |||
XM_006234514.1 | XP_006234576.1 | 11 | 1296 | |||
XM_006234516.1 | XP_006234578.1 | 11 | 1296 | |||
XM_006234518.1 | XP_006234580.1 | 11 | 1293 | |||
XM_006234519.1 | XP_006234581.1 | 11 | 1284 | |||
XM_006234520.1 | XP_006234582.1 | 11 | 1281 | |||
XM_006234521.1 | XP_006234583.1 | 11 | 1281 | |||
XM_006234522.1 | XP_006234584.1 | 11 | 1218 | |||
XM_006234523.1 | XP_006234585.1 | 12 | 1354 | |||
XM_006234524.2 | XP_006234586.1 | 12 | 1245 | |||
XM_006234526.2 | XP_006234588.1 | 12 | 1469 | |||
XM_006234527.2 | XP_006234589.1 | 12 | 1466 | |||
XM_008762008.1 | XP_008760230.1 | 13 | 2288 | |||
XM_008762009.1 | XP_008760231.1 | 13 | 1371 | |||
XM_008762010.1 | XP_008760232.1 | 12 | 2135 | |||
XM_008762011.1 | XP_008760233.1 | 11 | 1165 | |||
GenBank mRNA | BC098012.1 | - | 10 | 1155 | 7454358 | |
FQ221418.1 | - | 11 | 1287 | |||
FQ222063.1 | - | 11 | 1294 |
