siRNA Details
Gene Symbol: Trim44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013203.1 | NP_001013221.1 | 3 | 964 | ||
GenBank mRNA | AY551091.1 | - | 3 | 964 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_020267, BC098369, AJ249129, BC045602, AJ278191, and others.
Gene Ontology Categories:
