siRNA Details
Gene Symbol: Dnajc24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191853.1 | NP_001178782.1 | 3 | 272 | ||
XM_006234653.2 | XP_006234715.1 | 3 | 316 | |||
XM_006234654.2 | XP_006234716.1 | 3 | 305 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
