siRNA Details
Gene Symbol: Ccndbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013204.1 | NP_001013222.1 | 11 | 1004 | ||
XM_006234859.1 | XP_006234921.1 | 10 | 1052 | |||
GenBank mRNA | BC091256.1 | - | 11 | 1004 | 7375652 | |
FQ233751.1 | - | 11 | 1045 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
