siRNA Details
Gene Symbol: Shf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006234884.2 | XP_006234946.1 | 3 | 351 | ||
XM_006234885.2 | XP_006234947.1 | 4 | 492 | |||
XM_008762216.1 | XP_008760438.1 | 5 | 795 | |||
XM_008762217.1 | XP_008760439.1 | 4 | 492 | |||
XM_008762218.1 | XP_008760440.1 | 5 | 795 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
